Learn More
MESA LABS BOZEMAN MFG FACILITY MAGNA-AMPS 50/PK
SDP
Catalog No. NC9661445
Encompass
MagnaAmp,COLOR CHANGE AMPOULES FOR MONITORING STEAM STERILIZATION OF LIQUIDS,Process Steam sterilization of liquids ,spores:contain spores of Geobacillus stearothermophilus (Bacillus stearothermophilus),50/Pack
Catalog No. NC9661445 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. MA/5
MagnaAmp,COLOR CHANGE AMPOULES FOR MONITORING STEAM STERILIZATION OF LIQUIDS,Process Steam sterilization of liquids ,spores:contain spores of Geobacillus stearothermophilus (Bacillus stearothermophilus),50/Pack