Learn More
MESA LABS BOZEMAN MFG FACILITY MAGNA-AMPS MA/6 50/PK
SDP
Catalog No. NC9938309
Encompass
MagnaAmp,COLOR CHANGE AMPOULES FOR MONITORING STEAM STERILIZATION OF LIQUIDS,Process Steam sterilization of liquids ,spores:contain spores of Geobacillus stearothermophilus (Bacillus stearothermophilus),ATCC no.7953
Catalog No. NC9938309 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. MA/6
MagnaAmp,COLOR CHANGE AMPOULES FOR MONITORING STEAM STERILIZATION OF LIQUIDS,Process Steam sterilization of liquids ,spores:contain spores of Geobacillus stearothermophilus (Bacillus stearothermophilus),ATCC no.7953