Learn More
Roche Diagnostics MagNA Pure 96 Bacterial Lysis Buffer, 20 mL
Supplier:  Roche Diagnostics 06374921001
Encompass
The MagNA Pure 96 Bacterial Lysis Buffer is designed for: Lysis of different/various sample materials, stabilization of nucleic acids with lysates, and nucleic acid purification using the MagNA Pure 96 System.
Catalog No. NC1055129