Learn More
Roche Diagnostics MagNA Pure 96 DNA Tissue Lysis Buffer, 100 mL
Supplier:  Roche Diagnostics 06640702001
Encompass
The MagNA Pure 96 Tissue Lysis Buffer is designed for: Lysis of different types of tissues, Stabilization of DNA in tissue lysates, and Purification of genomic DNA from tissue sample lysates using the MagNA Pure 96 System.
Catalog No. NC1177614