Learn More
Roche Diagnostics MagNA Pure 96 External Lysis Buffer, 100 mL
Supplier:  Roche Diagnostics 06374913001
Encompass
The MagNA Pure 96 External Lysis Buffer is designed for: Lysis of different/various sample materials, Stabilization of nucleic acids with lysates, and Nucleic acid purification using the MagNA Pure 96 System.
Catalog No. NC1177612