Learn More
Roche Diagnostics MagNA Pure 96 Output Plate, 60 plates
Supplier:  Roche Diagnostics 06241611001
Encompass
MagNA Pure 96 disposables are nuclease free according to current Quality Control procedures. They are manufactured from material that is inert to chemical leaching at low and elevated temperatures, that could interfere with downstream applications.
Catalog No. NC1741499