Learn More
MSC CST/berger Magna-Trak MT200 Magnetic Locator, Ferrous Metal Detection, 4.6 m Depth, Digital Display, 1 ea
Catalog No. 5000510983
Encompass
The MT200 is a flux-gate magnetometer designed for the precise detection of ferrous metal objects. This instrument utilizes patented sensing coil technology to identify ferromagnetic fields, providing users with a clear peak audio response and visual signal indication. It is engineered for durability, featuring a water-resistant enclosure and a sensor pole that allows for submersion in water up to just under the plastic case.
  • Detects ferrous targets like manhole covers, survey markers, and iron pipes
  • LCD display with numeric signal, analog bar-graph, and power line alert
  • Adjustable volume and sensitivity controls via digital push buttons
  • Visual low battery status indicator
  • Operates on six AA batteries with up to 100 hours of intermittent life
  • Lightweight, balanced design
  • High-impact plastic housing
Catalog No. 50-005-10983 Supplier MSC Supplier No. 84334838
The MT200 is a flux-gate magnetometer designed for the precise detection of ferrous metal objects. This instrument utilizes patented sensing coil technology to identify ferromagnetic fields, providing users with a clear peak audio response and visual signal indication. It is engineered for durability, featuring a water-resistant enclosure and a sensor pole that allows for submersion in water up to just under the plastic case.
  • Detects ferrous targets like manhole covers, survey markers, and iron pipes
  • LCD display with numeric signal, analog bar-graph, and power line alert
  • Adjustable volume and sensitivity controls via digital push buttons
  • Visual low battery status indicator
  • Operates on six AA batteries with up to 100 hours of intermittent life
  • Lightweight, balanced design
  • High-impact plastic housing