Learn More
Description
Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.
MagnaBind Amine Derivatized Beads are supplied as an aqueous suspension of magnetic iron oxide beads coated with amine groups for covalent coupling of molecules using a variety of crosslinkers.
Features of MagnaBind Amine Derivatized Beads:
- Composition: Silanized iron oxide
- Magnetization: 25-35EMU/g
- Type of Magnetization: Superparamagnetic (no magnetic memory)
- Surface Area: >100m2/g
- Bead Size: 1-4μm diameter
- Settling Rate: 4% in 30 minutes
- Effective Density: 2.5g/mL
- Number of Beads: 1 x 108 beads/mg
- pH Stability: Aqueous solution, above pH 4.0
- Concentration: ∼50 mg/mL
Applications:
- Cell sorting using either positive or negative selection
- Protein purification or immunoassays using either direct or indirect methods
Specifications
Specifications
| Concentration | Slurry: ∼50 mg/mL |
| Content And Storage | Upon receipt store product at 4°C. Do not freeze product. Product is shipped at ambient temperature. |
| Description | MagnaBind Amine Derivatized Beads |
| Color | Brown |
| For Use With (Application) | Affinity Purification |
| Product Type | Amine Derivatized Bead |
| Product Line | MagnaBind |
| Quantity | 5 mL |
| Surface Functionality | Amine |
| Volume (Metric) | 5 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.