Learn More
Electron Microscopy Sciences Magnabrite™ 64 mm Diameter Base
SDP
Catalog No. 5027011
Encompass_Preferred
Made from solid grounded acrylic (not molded) and polished like a glass lens. Crystal clear and pre-focused. There are no bulbs or batteries needed. The 4x magnification Light-Gathering Magnifiers intensifying available light nearly four times and focusing it on the subject area. An "Infinite eyepoint" eliminates the need to keep the eye close to the magnifier so the object can be viewed from any distance, at any angle. It reduces eye strain and allows several people to view the same object at once.
Catalog No. 50-270-11 Supplier Electron Microscopy Sciences Supplier No. 7439364
Made from solid grounded acrylic (not molded) and polished like a glass lens. Crystal clear and pre-focused. There are no bulbs or batteries needed. The 4x magnification Light-Gathering Magnifiers intensifying available light nearly four times and focusing it on the subject area. An "Infinite eyepoint" eliminates the need to keep the eye close to the magnifier so the object can be viewed from any distance, at any angle. It reduces eye strain and allows several people to view the same object at once.