Learn More
Sentry Air Systems Model 300 Magnehelic Gauge
SDP
Catalog No. NC2831909
Encompass
The Magnehelic Gauge monitors the filter saturation for Model 300 units. This item displays the static pressure on the filter in inches of water column. For use with HEPA, ASHRAE, or ULPA filters only.
Catalog No. NC2831909 Supplier Sentry Air Systems Supplier No. SS300MAG
The Magnehelic Gauge monitors the filter saturation for Model 300 units. This item displays the static pressure on the filter in inches of water column. For use with HEPA, ASHRAE, or ULPA filters only.