Learn More
Medchemexpress LLC Magnesium, [(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl (3S,4S,21R)-9-ethenyl-14-ethyl-13-formyl | 519-62-0 | MFCD00079053 | 95.0% | 908.48 g/mol | C55H71MgN4O6+ | 1 MG
SDP
Catalog No. 5000282605
Encompass_Preferred
Chlorophyll b is a research-grade photosynthetic pigment and tetrapyrrole derivative used in plant biochemistry and antioxidant studies. It functions as a hydrogen donor and complements chlorophyll a in light-harvesting complexes, and is applied in experiments on photosynthesis, oxidative stress, and DNA damage modulation.
  • Tetrapyrrole pigment for research use.
  • Acts as a hydrogen donor and antioxidant.
  • Supports studies of photosynthesis and light-harvesting complexes.
  • Has been reported to reduce cisplatin-induced DNA damage and oxidative stress in models.
  • Available in small pack sizes suitable for analytical and biochemical work.
Catalog No. 50-002-82605 Supplier Medchemexpress LLC Supplier No. HYW1277091MG
Chlorophyll b is a research-grade photosynthetic pigment and tetrapyrrole derivative used in plant biochemistry and antioxidant studies. It functions as a hydrogen donor and complements chlorophyll a in light-harvesting complexes, and is applied in experiments on photosynthesis, oxidative stress, and DNA damage modulation.
  • Tetrapyrrole pigment for research use.
  • Acts as a hydrogen donor and antioxidant.
  • Supports studies of photosynthesis and light-harvesting complexes.
  • Has been reported to reduce cisplatin-induced DNA damage and oxidative stress in models.
  • Available in small pack sizes suitable for analytical and biochemical work.