Learn More
Alkali Scientific Magnesium Acetate Tetrahydrate, 1 Kg
SDP
Catalog No. 5000659704
Encompass_Preferred
Magnesium Acetate Tetrahydrate, laboratory grade, is a chemical reagent available in a 1kg quantity, used in a variety of laboratory and industrial applications. This compound is utilized in biochemical experiments, particularly in the preparation of buffers and reagents for enzyme assays, protein research, and DNA/RNA analysis. Magnesium acetate serves as a source of magnesium ions, which are essential cofactors for numerous enzymatic reactions, including those involved in DNA replication and repair. It is also employed in the synthesis of other magnesium compounds and is used in laboratory studies that require precise ionic control in biochemical systems.
Catalog No. 50-006-59704 Supplier Alkali Scientific Supplier No. GNE002CP
Magnesium Acetate Tetrahydrate, laboratory grade, is a chemical reagent available in a 1kg quantity, used in a variety of laboratory and industrial applications. This compound is utilized in biochemical experiments, particularly in the preparation of buffers and reagents for enzyme assays, protein research, and DNA/RNA analysis. Magnesium acetate serves as a source of magnesium ions, which are essential cofactors for numerous enzymatic reactions, including those involved in DNA replication and repair. It is also employed in the synthesis of other magnesium compounds and is used in laboratory studies that require precise ionic control in biochemical systems.