Learn More
Medchemexpress LLC Magnesium acetate tetrahydrate | 16674-78-5 | MFCD00149214 | 99.5% | 214.45 g/mol | C4H14MgO8 | 100g
SDP
Catalog No. 5000269772
Encompass_Preferred
Magnesium acetate tetrahydrate is a carboxylic acid and short-chain fatty acid (SCFAs) Magnesium acetate tetrahydrate activates AMPK increases ROS cleaved caspase 9 PPAR downregulates SREBP-1c ChREBP expression Magnesium acetate tetrahydrate exhibits antifungal activity against Saccharomyces cerevisiae W303-1A Magnesium acetate tetrahydrate regulates energy metabolism Magnesium acetate tetrahydrate has anticancer activity against gastric cancer Magnesium acetate tetrahydrate induces writhing reaction and ulcerative colitis Magnesium acetate tetrahydrate can be used in the researches for gastric cancer ulcerative colitis hepatic steatosis and pain[1][2][3][4][5][6][7][8][9]
Catalog No. 50-002-69772 Supplier Medchemexpress LLC Supplier No. HYY0319G100G
Magnesium acetate tetrahydrate is a carboxylic acid and short-chain fatty acid (SCFAs) Magnesium acetate tetrahydrate activates AMPK increases ROS cleaved caspase 9 PPAR downregulates SREBP-1c ChREBP expression Magnesium acetate tetrahydrate exhibits antifungal activity against Saccharomyces cerevisiae W303-1A Magnesium acetate tetrahydrate regulates energy metabolism Magnesium acetate tetrahydrate has anticancer activity against gastric cancer Magnesium acetate tetrahydrate induces writhing reaction and ulcerative colitis Magnesium acetate tetrahydrate can be used in the researches for gastric cancer ulcerative colitis hepatic steatosis and pain[1][2][3][4][5][6][7][8][9]