Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium acetate tetrahydrate, 16674-78-5, MFCD00149214, 50 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M566150G
Encompass
Linear Formula: (CH3COO)2Mg · 4H2O, Molecular Weight: 214.45, for molecular biology, ≥99%, Synonym: Acetic acid magnesium salt.
Catalog No. NC1161060