Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium bromide tetrahydrofuran complex, 97%, Thermo Scientific™

Catalog No. AC387751000
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC387751000 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC387751000 Supplier Thermo Scientific Chemicals Supplier No. 387751000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63994-15-0
Quantity 100g
Assay Percent Range 97%
Molecular Formula C8H16Br2MgO2
MDL Number MFCD09751420
Synonym magnesiumbromidetetrahydrofurancomplex
InChI Key XBPFBACMBZKMJI-UHFFFAOYSA-L
SMILES C1CCOC1.C1CCOC1.[Mg+2].[Br-].[Br-]
IUPAC Name magnesium;oxolane;dibromide
Molecular Weight (g/mol) 328.327
PubChem CID 60160940
Formula Weight 328.33
Percent Purity ≥96.0%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Magnesium bromide tetrahydrofuran complex
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.