Learn More
Strem, An Ascensus Company CAS# 39409-82-0. 500g. Magnesium carbonate, basic pentahydrate, 99%. MFCD00149784
SDP
Catalog No. 5090113934
Encompass
CAS# 39409-82-0. 500g. Magnesium carbonate, basic pentahydrate, 99%. MFCD00149784. Molecular Weight: 395.57 (485.69). Molecular Formula: (MgCO3)4·;Mg(OH)2·;5H2O. Color/form: white pwdr. Strem# 93-1220. http://www.strem.com/catalog/v/93-1220/
Catalog No. 50-901-13934 Supplier Strem, An Ascensus Company Supplier No. 931220500G
CAS# 39409-82-0. 500g. Magnesium carbonate, basic pentahydrate, 99%. MFCD00149784. Molecular Weight: 395.57 (485.69). Molecular Formula: (MgCO3)4·;Mg(OH)2·;5H2O. Color/form: white pwdr. Strem# 93-1220. http://www.strem.com/catalog/v/93-1220/