Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium carbonate basic, heavy, extra pure

Catalog No. AC413400100
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000 | CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 10 kg
Linear Formula MgCO3
Merck Index 15,5724
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
Formula Weight 84.31
Percent Purity 40 to 45% (MgO)
Grade Extra Pure
Packaging Plastic drum
Chemical Name or Material Magnesium carbonate basic, heavy
Show More Show Less
EINECSNumber 208-915-9
RTECSNumber OM2470000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.