Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium carbonate, for biochemistry, specified according to the requirements of USP

Catalog No. AC439132500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439132500 250 g Plastic bottle
AC439130010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC439132500 Supplier Thermo Scientific Chemicals Supplier No. 439132500
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 546-93-0 | CMgO3 | 84.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-93-0
Molecular Formula CMgO3
Molecular Weight (g/mol) 84.31
MDL Number MFCD00064632
InChI Key ZLNQQNXFFQJAID-UHFFFAOYSA-L
Synonym magnesium carbonate, magnesite, kimboshi, magmaster, apolda, destab, magfy, magnesite dust, carbonic acid, magnesium salt, carbonate magnesium
PubChem CID 11029
ChEBI CHEBI:31793
IUPAC Name magnesium(2+) carbonate
SMILES [Mg++].[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Linear Formula MgCO3
Solubility Information Solubility in water: slightly soluble
Formula Weight 84.31
Percent Purity 40.0 to 43.5% (MgO)
Grade Biochemical
Identification conform
Packaging Plastic bottle
Chemical Name or Material Magnesium carbonate
Show More Show Less
EINECSNumber 208-915-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.