Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Magnesium carbonate hydroxide pentahydrate

Catalog No. 503124396 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-4396 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-312-4396 Supplier Sigma Aldrich Supplier No. M5671500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Magnesium carbonate hydroxide pentahydrate

Specifications

CAS 56378-72-4
Molecular Formula C4Mg4O12 -+ H2MgO2 -+ 5H2O
Linear Formula (MgCO3)4 -+ Mg(OH)2 -+ 5H2O
Quantity 500 g
MDL Number MFCD07371358
Synonym Basic magnesium carbonate
Molecular Weight (g/mol) 485.65
Percent Purity 40-43.5%NaOH basis (titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.