Learn More
Strem, An Ascensus Company CAS# 7439-95-4. 1kg. Magnesium chips (99+%). MFCD00085308
SDP
Catalog No. 5090113995
Encompass
CAS# 7439-95-4. 1kg. Magnesium chips (99+%). MFCD00085308. Molecular Weight: 24.32. Molecular Formula: Mg. Color/form: -4 + 18 mesh Strem# 93-1299. http://www.strem.com/catalog/v/93-1299/
Catalog No. 50-901-13995 Supplier Strem, An Ascensus Company Supplier No. 9312991000G
CAS# 7439-95-4. 1kg. Magnesium chips (99+%). MFCD00085308. Molecular Weight: 24.32. Molecular Formula: Mg. Color/form: -4 + 18 mesh Strem# 93-1299. http://www.strem.com/catalog/v/93-1299/