Learn More
Hach Company Magnesium Chloride Digital Titrator Cartridge, 0.080 M
Catalog No. 501995197 Shop All Hach Company Products
Encompass
For chelant determination by Digital Titrator titration. 0.080 M. Digits x 0.1 = mg/L chelant (as mg/L CaCO₃) in 100 mL sample.
Catalog No. 50-199-5197 Supplier Hach Company Supplier No. 2062501
For chelant determination by Digital Titrator titration. 0.080 M. Digits x 0.1 = mg/L chelant (as mg/L CaCO₃) in 100 mL sample.