Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride, 1M aq. soln.

Catalog No. AAJ62411AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62411AE 100 mL
AAJ62411AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62411AE Supplier Thermo Scientific Chemicals Supplier No. J62411AE
Add to Cart
Edge

missing translation for 'validMainframeMsg'

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

Solubility
Soluble in water (542 mg/ml at 20°C), alcohol, methanol (15.5 g/100g at 0°C), and ethanol (3.61 g/100g at 0°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Very hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Physical Form Liquid
Quantity 100 mL
Linear Formula MgCl2
Solubility Information Soluble in water (542mg/ml at 20°C),alcohol,methanol (15.5g/100g at 0°C),and ethanol (3.61g/100g at 0°C).
IUPAC Name magnesium(2+) dichloride
Formula Weight 95.22
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Magnesium chloride
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.