Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, 500 g, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S001064 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001064 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001064 Supplier Aldon Corporation Supplier No. IS22011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.31

Specifications

Physical Form Powder, Needles or Crystals
Quantity 500 g
Percent Purity 100%
Grade ACS
Packaging HDPE Bottle
Chemical Name or Material Magnesium chloride, hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.