Learn More
Molecular Dimensions 5M Magnesium chloride hexahydrate, 7791-18-6, MFCD00149781, 250mL
SDP
Catalog No. NC0630364
Encompass
Molecular Formula Cl2Mg.6H2O, Molecular Weight 203.300; 5M Magnesium chloride hexahydrate solution. Made with ultra-purified water and filter sterilized.
Catalog No. NC0630364 Supplier Molecular Dimensions Supplier No. MD225047
Molecular Formula Cl2Mg.6H2O, Molecular Weight 203.300; 5M Magnesium chloride hexahydrate solution. Made with ultra-purified water and filter sterilized.