Learn More
Medchemexpress LLC Magnesium chloride | 7786-30-3 | MFCD00011106 | 97.0% | 95.21 | MgCl2 | 5g
SDP
Catalog No. 5000267502
Encompass_Preferred
Magnesium chloride for insect cell culture 97% (Magnogene for insect cell culture 97%) is a magnesium chloride reagent specifically used for insect cell culture with a purity of 97% Magnesium chloride for insect cell culture 97% plays an important role in maintaining osmotic pressure regulating enzyme activity stabilizing cell structure and assisting the buffering system[1]
Catalog No. 50-002-67502 Supplier Medchemexpress LLC Supplier No. HYY1110I5G
Magnesium chloride for insect cell culture 97% (Magnogene for insect cell culture 97%) is a magnesium chloride reagent specifically used for insect cell culture with a purity of 97% Magnesium chloride for insect cell culture 97% plays an important role in maintaining osmotic pressure regulating enzyme activity stabilizing cell structure and assisting the buffering system[1]