Learn More
Medchemexpress LLC Magnesium chloride anhydrous | 7786-30-3 | MFCD00011106 | 97.0% | 95.21 g·mol⁻¹ | MgCl2 | 1g
SDP
Catalog No. 5000267988
Encompass_Preferred
Magnesium chloride anhydrous suitable for insect cell culture is used for preparing Tris buffer and also for cell culture
Catalog No. 50-002-67988 Supplier Medchemexpress LLC Supplier No. HYY1110H1G
Magnesium chloride anhydrous suitable for insect cell culture is used for preparing Tris buffer and also for cell culture