Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium chloride hexahyd100G
Catalog No. 502897076
Encompass
Magnesium chloride hexahydrate (MgCl2A6H2O) is an inorganic salt that can be generated from the acid leach SOLUTIONon of magnesite tailings. It is a potential phase change material (PCM) for storing thermal energy in solar cookers. The result of altering poly (vinyl alcohol)(PVA) films with MgCl2A6H2O has been investigated. The ability of MgCl2A6H2O to reduce the flammability of cotton fabrics has been studied.
Catalog No. 50-289-7076 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M9272100G
Magnesium chloride hexahydrate (MgCl2A6H2O) is an inorganic salt that can be generated from the acid leach SOLUTIONon of magnesite tailings. It is a potential phase change material (PCM) for storing thermal energy in solar cookers. The result of altering poly (vinyl alcohol)(PVA) films with MgCl2A6H2O has been investigated. The ability of MgCl2A6H2O to reduce the flammability of cotton fabrics has been studied.