Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate
SCI_ED

Catalog No. S93290 Shop All Fisherbrand Products
Change view
Click to view available options
Lab Grade - Crystal; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S93290 Lab Grade - Crystal; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S93290

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color White
CAS 7791-18-6
Quantity 500 g
Molecular Formula Cl2H12MgO6
Linear Formula MgCl2·6H2O
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Formula Weight 203.31
Packaging Lab Grade - Crystal
Chemical Name or Material Magnesium Chloride
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.