Learn More
Medchemexpress LLC Magnesium chloride hexahydrate | 7791-18-6 | MFCD00149781 | 100.4% | 203.30 g/mol | MgCl2·6H2O | 500 MG
SDP
Catalog No. 5000300955
Encompass_Preferred
Magnesium chloride hexahydrate (MgCl2·6H2O) is a water-soluble, white to off-white solid used to supply magnesium ions in cell culture and biochemical assays. It provides Mg2+ as an essential cofactor for many enzymes and is suitable for media supplementation and laboratory reagent use.
  • Provides soluble magnesium ions for cell culture and enzymatic reactions.
  • Acts as a cofactor to support enzyme activity.
  • White to off-white solid with high solubility in water.
  • Available in laboratory-sized packs for research use.
  • Quality documentation available including datasheet, SDS, and batch COA.
Catalog No. 50-003-00955 Supplier Medchemexpress LLC Supplier No. HYW019981C500MG
Magnesium chloride hexahydrate (MgCl2·6H2O) is a water-soluble, white to off-white solid used to supply magnesium ions in cell culture and biochemical assays. It provides Mg2+ as an essential cofactor for many enzymes and is suitable for media supplementation and laboratory reagent use.
  • Provides soluble magnesium ions for cell culture and enzymatic reactions.
  • Acts as a cofactor to support enzyme activity.
  • White to off-white solid with high solubility in water.
  • Available in laboratory-sized packs for research use.
  • Quality documentation available including datasheet, SDS, and batch COA.