Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Magnesium chloride hexahydrate | 7791-18-6 | MFCD00149781 | 99.4% | 203.30 g·mol⁻¹ | MgCl2·6H2O | 10 G
SDP

Catalog No. 5000301521
Encompass_Preferred

Magnesium chloride hexahydrate (MgCl2·6H2O) is a reagent-grade inorganic salt used as a source of magnesium ions in molecular biology applications. It serves as a cofactor for many enzymes and can enhance enzymatic activity in buffers and reactions; supplied as a white to off-white crystalline solid for research use only.

  • Provides magnesium ions for enzyme cofactors and buffer formulation.
  • High purity: 99.4%.
  • Molecular weight: 203.30 g·mol⁻¹.
  • Available in laboratory pack sizes (10 g, 25 g, 50 g).
  • Documentation available: datasheet, certificate of analysis, and SDS.

Catalog No. 50-003-01521 Supplier Medchemexpress LLC Supplier No. HYW019981B10G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnesium chloride hexahydrate (MgCl2·6H2O) is a reagent-grade inorganic salt used as a source of magnesium ions in molecular biology applications. It serves as a cofactor for many enzymes and can enhance enzymatic activity in buffers and reactions; supplied as a white to off-white crystalline solid for research use only.

  • Provides magnesium ions for enzyme cofactors and buffer formulation.
  • High purity: 99.4%.
  • Molecular weight: 203.30 g·mol⁻¹.
  • Available in laboratory pack sizes (10 g, 25 g, 50 g).
  • Documentation available: datasheet, certificate of analysis, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.