Learn More
Medchemexpress LLC Magnesium chloride hexahydrate | 7791-18-6 | MFCD00149781 | 99.4% | 203.30 g/mol | MgCl2·6H2O | 25 G
SDP
Catalog No. 5000303035
Encompass_Preferred
Magnesium chloride hexahydrate is a high-purity inorganic salt supplied as the hexahydrate form (MgCl2·6H2O). It provides a reliable source of magnesium ions for molecular biology and biochemical applications, serving as an enzyme cofactor, buffer component, and reagent for assays requiring controlled Mg2+ concentrations.
  • High purity 99.4%
  • Provides Mg2+ ions as an enzyme cofactor
  • Suitable for molecular biology and biochemical assays
  • Water-soluble crystalline hexahydrate form
  • Stable under normal laboratory storage conditions
  • Convenient small-scale laboratory packaging
Catalog No. 50-003-03035 Supplier Medchemexpress LLC Supplier No. 25GHYW019981B
Magnesium chloride hexahydrate is a high-purity inorganic salt supplied as the hexahydrate form (MgCl2·6H2O). It provides a reliable source of magnesium ions for molecular biology and biochemical applications, serving as an enzyme cofactor, buffer component, and reagent for assays requiring controlled Mg2+ concentrations.
  • High purity 99.4%
  • Provides Mg2+ ions as an enzyme cofactor
  • Suitable for molecular biology and biochemical assays
  • Water-soluble crystalline hexahydrate form
  • Stable under normal laboratory storage conditions
  • Convenient small-scale laboratory packaging