Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride hexahydrate, >98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1228836

Quantity 500g

 View more versions of this product

Catalog No. AA1228836


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7791-18-6
>98%
203.30 (95.22anhydrous)
Magnesium chloride hexahydrate
Flakes
MgCl2·6H2O
MFCD00149781

Specifications

7791-18-6
>98%
White flakes
MFCD00149781
Odorless
500g
Flakes
MgCl2·6H2O
203.30 (95.22anhydrous)
1.57g/cm3 at 20°C
White
Magnesium chloride hexahydrate

Safety and Handling

SDS & Safety Documents

EINECSNumber : 232-094-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit