Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, Hexahydrate, ACS, 99.0 to 101.0%, MilliporeSigma™

Catalog No. M1058331000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1058331000 1 kg
M1058330250 250 g
M1058335000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1058331000 Supplier MilliporeSigma Supplier No. 1.05833.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym Magnesium dichloride
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Color White to Gray White
Physical Form Solid
Quantity 1 kg
Assay Percent Range 99.0 to 101.0%
Linear Formula MgCl2 · 6H2O
Formula Weight 23.3
Grade ACS/EP/ISO/Reagent
Density 1.57g/cm3 (20°C)
Chemical Name or Material Magnesium Chloride, Hexahydrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.