Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCM0030500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCM0030500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCM0030500 Supplier Ricca Chemical Company Supplier No. RDCM0030500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

CAS Min % 98.0
Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100.0
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Percent Purity 99 to 102%
Grade ACS
Chemical Name or Material Magnesium Chloride, Hexahydrate
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.