Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP, JPC, FCC, bioCERTIFIED™, 2.5 kg, Spectrum Chemical
SDP

Catalog No. 18615626 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-615-626 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-615-626 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M91002.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity magnesium chloride hexahydrate suitable for critical laboratory and industrial applications.

100% Tested. 100% Transparent. 100% Trusted.

Spectrum Chemical bioCERTIFIED™ products are uniquely suited for bioprocessing by biopharmaceutical manufacturers. From drug discovery to pilot and scale-up to production, bioCERTIFIED™ products help simplify and standardize bioprocessing by ensuring consistent raw material quality. All bioCERTIFIED™ materials and finished goods are subject to in-house laboratory testing and analysis in cGMP-compliant facilities. This includes expanded testing for bioburden, endotoxin and elemental impurities.

  • Extensive product breadth and packaging flexibility for upstream and downstream bioprocessing
  • Expanded laboratory testing, certifications and documentation including additional lot testing for bioburden, endotoxins and elemental impurities
  • Comprehensive regulatory and scientific documentation to help meet global quality, regulatory and compliance requirements
  • Manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA-registered and inspected facilities
  • Change control, lot traceability and supply chain transparency to eliminate unpredictability

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.3

Specifications

Physical Form Crystals and/or Chunks
Quantity 2.5 kg
Packaging Poly Bottle
Percent Purity 98 to 101%
Grade BP/EP/FCC/USP/bioCERTIFIED
Chemical Name or Material Magnesium Chloride, Hexahydrate, Crystal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.