Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1058339025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1058339025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1058339025 Supplier MilliporeSigma Supplier No. 1058339025
Add to Cart
Add to Cart

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym Magnesium Dichloride
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C (decomposition)
pH 4.5 to 7.0 (50g/L, H2O, 20°C)
Quantity 25 kg
Solubility Information 1670g/L
Formula Weight 203.3
Percent Purity 99.0 to 101.0%
Grade ACS
Packaging Fiber Carton
Density 1.57 g/3 (20 °C)
EINECSNumber 232-094-6
RTECSNumber OM2975000
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.