Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate, MACCO

Catalog No. 502985581
Encompass
Catalog No. 50-298-5581 Supplier Thermo Scientific Production Chemicals and Services Supplier No. M51203060120
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.30 g/mol

Specifications

Physical Form Crystalline Powder
Quantity 12 kg
Percent Purity 99 to 100%
Grade USP
Packaging 12 kg poly drum Curtec + 2 x PE liner inside
Chemical Name or Material Magnesium chloride hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.