Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate, MP Biomedicals™

Catalog No. MP021914215 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914215 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021914215 Supplier MP Biomedicals, Inc Supplier No. 02191421.5
Add to Cart
Add to Cart

Used to prepare renaturation buffer. Magnesium Chloride Hexahydrate, MP Biomedicals is an inorganic salt that can be generated from the acid leach solution of magnesite tailings. It is a potential phase change material (PCM) for storing thermal energy in

Magnesium chloride hexahydrate (MgCl2.6H2O) has been used to prepare renaturation buffer that is employed for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes. It may be used as a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones. The result of altering poly(vinyl alcohol) (PVA) films with magnesium chloride hexahydrate has been investigated. Its ability to reduce the flammability of cotton fabrics has also been studied.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym Magnesium dichloride hexahydrate, Magnesium chloride hydrate, Magnesium chloride
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystals
Quantity 500 g
Solubility Information Soluble in water (500mg/mL), yielding a clear, colorless solution.
Formula Weight 203.3
Percent Purity 99 to 102%
Grade ACS Reagent
Recommended Storage Room Temperature (15-30°C), dessicate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.