Learn More
New England Biolabs, Inc. Magnesium Chloride (MgCl2) Solution – 6 ml (4 x 1.5 ml)
SDP
Catalog No. 504289
Encompass
25 mM MgCl2 for use in conjunction with polymerase reaction buffers.
Catalog No. 50-428-9 Supplier New England Biolabs, Inc. Supplier No. B9021S
25 mM MgCl2 for use in conjunction with polymerase reaction buffers.