Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium Bromide, 99+%

Catalog No. AC227160101
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227160101 10 kg Plastic drum
AC227160100 10 g Glass Bottle
AC227160250 25 g Plastic Bottle
AC227161000 100 g Plastic Bottle
AC227165000 500 g Plastic Bottle
AC227160025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AC227160101 Supplier Thermo Scientific Chemicals Supplier No. 227160101
Only null left
Add to Cart

CAS: 57-09-0 | C19H42BrN | 364.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
PubChem CID 5974
ChEBI CHEBI:3567
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Physical Form Crystalline Powder
Melting Point 230.0°C
Decomposition Information >230°C
Quantity 10 kg
Flash Point 244°C
Infrared Spectrum Authentic
Packaging Plastic drum
Assay Percent Range 99+%
Linear Formula CH3(CH2)15N(CH3)3Br
Beilstein 04, IV, 819
Fieser 15,77
Merck Index 15, 2024
Synonym Cetrimonium bromide, Cetyltrimethylammonium bromide, CTABr
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone
IUPAC Name hexadecyl(trimethyl)azanium;bromide
Formula Weight 364.45
Percent Purity ≥99%
Chemical Name or Material Hexadecyltrimethylammonium bromide
Show More Show Less
Hazard Category Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.