Learn More
Crescent Chemical Co Inc MAGNESIUM CHLORIDE SOLN 1 M
SDP
Catalog No. 50248457
Encompass_Preferred
Magnesium chloride, solution 1 M molecular biology grade DNase/RNase not detected. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39772.01
Catalog No. 50-248-457 Supplier Crescent Chemical Co Inc Supplier No. SE39772.01
Magnesium chloride, solution 1 M molecular biology grade DNase/RNase not detected. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39772.01