Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium chloride solution, 7786-30-3, MFCD00011106, 100mL
Catalog No. 501657248
Encompass
Linear Formula MgCl2, Molecular Weight 95.21, for molecular biology, 1.00 M±0.01 M
Catalog No. 50-165-7248 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M1028100ML
Linear Formula MgCl2, Molecular Weight 95.21, for molecular biology, 1.00 M±0.01 M