Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride, ultra dry, 99.9% (metals basis)

Catalog No. AA3570709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3570709 10 g
AA3570718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3570709 Supplier Thermo Scientific Chemicals Supplier No. 03570709
Add to Cart

Widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. | CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

It is widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. It is an important coagulant, an elution buffer for antibody affinity column purifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
IUPAC Name magnesium;dichloride
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Melting Point 714°C
Physical Form Beads
Quantity 10 g
Linear Formula MgCl2
Solubility Information Soluble in water,methanol,ethanol and alcohol.
Formula Weight 95.22
Percent Purity 99.9%
Packaging Ampouled under argon
Sensitivity Hygroscopic
Specific Gravity 2.32
Density 2.32
Chemical Name or Material Magnesium chloride
Show More Show Less
EINECSNumber 232-094-6
RTECSNumber OM2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.