Learn More
Magnesium chloride, ultra dry, 99.99% (metals basis)
Description
Magnesium chloride is generally used as a Lewis acid catalyst. It is used as a flocculating agent, analytical reagent, anesthetic for cephalopods, and also in various magnesium cements, and ceramics. It is used for dust control, and road stabilization. It is used in fire extinguishers, polymerase chain reaction (PCR) and hydrogen storage. It is a precursor for magnesium metal and other magnesium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Melting Point | 714°C |
| Color | White to Yellow |
| Physical Form | Beads |
| Quantity | 50 g |
| Linear Formula | MgCl2 |
| Merck Index | 14,5662 |
| Solubility Information | Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine. |
| IUPAC Name | magnesium;dichloride |
| Formula Weight | 95.22 |
| Percent Purity | 99.99% |
| Show More |
Safety and Handling
| EINECSNumber | 232-094-6 |
| RTECSNumber | OM2800000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.