Learn More
Medchemexpress LLC Magnesium citrate 500mg
SDP
Catalog No. 5000204953
Encompass_Preferred
Magnesium citrate (Trimagnesium dicitrate) is an orally active magnesium-containing compound that can cross the blood-brain barrier (BBB) Magnesium citrate can be used as an intestinal cleanser and is studied for its application in laxatives Magnesium citrate can also increase the pain threshold and is studied for pain-relief purposes[1][2]
Catalog No. 50-002-04953 Supplier Medchemexpress LLC Supplier No. HYW105835500MG
Magnesium citrate (Trimagnesium dicitrate) is an orally active magnesium-containing compound that can cross the blood-brain barrier (BBB) Magnesium citrate can be used as an intestinal cleanser and is studied for its application in laxatives Magnesium citrate can also increase the pain threshold and is studied for pain-relief purposes[1][2]