Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Citrate Tribasic, Spectrum™ Chemical
SDP

Catalog No. 18621003 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-003 50 kg
18-620-999 125 g
18-621-000 500 g
18-621-001 2.5 kg
18-621-002 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-621-003 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M103250KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Magnesium Citrate Tribasic Hydrate

Chemical Identifiers

CAS 3344-18-1

Specifications

Physical Form Granular/Lump
Quantity 50 kg
Molecular Weight (g/mol) 451.12
Percent Purity 14.5 to 16.4 %
Grade Not Available
Packaging Poly Bottle
Chemical Name or Material Magnesium Citrate Tribasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.