Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Di Citrate, MP Biomedicals

Catalog No. ICN212256 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212256 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212256 Supplier MP Biomedicals, Inc Supplier No. 0521225680
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3344-18-1
Molecular Formula C12H12Mg3O15
Molecular Weight (g/mol) 469.128
InChI Key HZTVQLXIRHEMMB-UHFFFAOYSA-H
Synonym trimagnesium 2+ dicitrate hydrate
PubChem CID 122198783
IUPAC Name trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2]

Specifications

Quantity 100 g
Formula Weight 703.36
Chemical Name or Material Magnesium Di Citrate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.