Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium ethoxide, 97+%

Catalog No. AC397471000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397471000 100 g Glass bottle
AC397470050 5 g Glass bottle
AC397475000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC397471000 Supplier Thermo Scientific Chemicals Supplier No. 397471000
Only null left
Add to Cart

CAS: 2414-98-4 | C4H10MgO2 | 114.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2414-98-4
Molecular Formula C4H10MgO2
Molecular Weight (g/mol) 114.43
MDL Number MFCD00009060
InChI Key XDKQUSKHRIUJEO-UHFFFAOYSA-N
Synonym magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
PubChem CID 164963
IUPAC Name magnesium;ethanolate
SMILES CC[O-].CC[O-].[Mg+2]

Specifications

Melting Point 270.0°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3CH2O)2Mg
Quantity 100 g
Solubility Information Solubility in water: reacts
Formula Weight 114.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Magnesium ethoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Self-heating; may catch fire.
Causes serious eye irritation.
Reacts violently with water.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
If
EINECSNumber 219-323-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.