Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Magnesium fluoride | 7783-40-6 | MFCD00011108 | 98.0% | 62.30 g/mol | MgF2 | 25 G
SDP

Catalog No. 5000303077
Encompass_Preferred

Magnesium fluoride (MgF2) is an inorganic fluoride salt supplied as a white to off-white solid for laboratory research. It is used as a reagent and biochemical assay material in inorganic synthesis, optics, and analytical applications. The product is provided with assay information and supporting documentation for research use.

  • White to off-white solid appearance.
  • Assay: 98%.
  • Chemical formula MgF2; molecular weight 62.30 g/mol.
  • Available pack sizes: 25 g, 100 g, larger quantities by quote.
  • Storage: 4°C, sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • SDS and certificate of analysis available.

Catalog No. 50-003-03077 Supplier Medchemexpress LLC Supplier No. 25GHYW019922
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnesium fluoride (MgF2) is an inorganic fluoride salt supplied as a white to off-white solid for laboratory research. It is used as a reagent and biochemical assay material in inorganic synthesis, optics, and analytical applications. The product is provided with assay information and supporting documentation for research use.

  • White to off-white solid appearance.
  • Assay: 98%.
  • Chemical formula MgF2; molecular weight 62.30 g/mol.
  • Available pack sizes: 25 g, 100 g, larger quantities by quote.
  • Storage: 4°C, sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • SDS and certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.