Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium formate dihydrate, 6150-82-9, MFCD00150512, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 00793100GF
Encompass
Empirical Formula (Hill Notation): C2H2MgO4 · 2H2O, Molecular Weight: 150.37, BioXtra, ≥98.0% (RT), Synonym: Formic acid magnesium salt. Magnesium formate dehydrate, or formic acid magnesium salt, has been used in studies to develop a multiresidue method for determining pesticides in fruits and vegetables, as well as for the simultaneous determination of tropane alkaloids and glycoalkaloids in grains and seeds.
Catalog No. NC1125284